Quartz Wool
Thermo Scientific Chemicals
Quartz Wool
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ACR451040500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV(1 M in water)
UVat 250 nm A: =<0.03
UVat 280 nm A: =<0.02
Appearance (Color)White