2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ACR450182500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min