tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number ACR447010025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7439-88-5
IUPAC Nameiridium
Molecular FormulaIr
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES[Ir]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA≥99.8% (metals basis)
Appearance (Color)Gray