Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR44436025025 LMetal drum
Catalog number ACR444360250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Chemical Identifiers
CAS34071-95-9
IUPAC Name2,5-dioxopyrrolidin-1-yl 3-(4-hydroxyphenyl)propanoate
Molecular FormulaC13H13NO5
InChI KeyKYRUKRFVOACELK-UHFFFAOYSA-N
SMILESOC1=CC=C(CCC(=O)ON2C(=O)CCC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥94.0%
Proton NMRConforms
Appearance (Color)White to off-white