N-Butylimidazole, 98%, Thermo Scientific Chemicals
N-Butylimidazole, 98%, Thermo Scientific Chemicals
N-Butylimidazole, 98%, Thermo Scientific Chemicals
N-Butylimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Butylimidazole, 98%, Thermo Scientific Chemicals
CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR442770250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Butylimidazole
CAS4316-42-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat