Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Thermo Scientific Chemicals
Trioctylphosphine, 90%, technical grade, AcroSeal™
CAS: 4731-53-7 | C24H51P | 370.64 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR436461000100 mLAcroSeal™ Glass Bottle
Catalog number ACR436461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Clarity of alcohol solutionPasses test
Visual transition intervalFrom pH 8.0 (colorless) to pH 10 (red)