n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
n-Nonane, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
n-Nonane, 99%, anhydrous, AcroSeal™
CAS: 111-84-2 | C9H20 | 128.26 g/mol
Have Questions?
Catalog NumberdimQuantityPackaging
ACR435631000
Catalog number ACR435631000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS13106-76-8
IUPAC Namediammonium dioxomolybdenumbis(olate)
Molecular FormulaH8MoN2O4
InChI KeyAPUPEJJSWDHEBO-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (special)?2.5 % of Theoretical value (Mo)
FormCrystalline powder and/or chunks
Total Metal Impurities≤0.01% (100 ppm)