Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Thermo Scientific Chemicals
Ethyl 2-cyanoacrylate
CAS: 7085-85-0 | C6H7NO2 | 125.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4344500505 gGlass bottle
Catalog number ACR434450050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS369-35-7
IUPAC Name2-fluoro-4-nitroaniline
Molecular FormulaC6H5FN2O2
InChI KeyLETNCFZQCNCACQ-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%