Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Thermo Scientific Chemicals
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol
Have Questions?
Catalog NumberQuantity
ACR427861000100 mL
Catalog number ACR427861000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS657-27-2
IUPAC Name(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-JEDNCBNOSA-N
SMILESCl.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals (as Pb)≤10ppm
CommentEndotoxins: ≤6.0EU/g
Impurity contentArsenic as AS2O3: ≤1ppm
Impurity contentIron, Fe: ≤10ppm
Impurity contentSulphate, SO4: ≤0.02% (≤200ppm)