Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR424535000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7758-89-6
IUPAC Nameλ¹-copper(1+) chloride
Molecular FormulaClCu
InChI KeyOXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES[Cl-].[Cu+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light green
Iron (Fe)=<50 ppm
Titration with KMnO4>=90.0 %
Calcium (Ca)=<100 ppm
Appearance (Form)Crystalline powder