Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number ACR424045000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2373-79-7
IUPAC Name(2E)-3-(4-ethoxyphenyl)prop-2-enoate
Molecular FormulaC11H11O3
InChI KeyDZLOUWYGNATKKZ-VMPITWQZSA-M
SMILESCCOC1=CC=C(C=CC([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)194.0-200.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%