Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR423485000500 gPlastic bottle
Catalog number ACR423485000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS112-36-7
IUPAC Name1-ethoxy-2-(2-ethoxyethoxy)ethane
Molecular FormulaC8H18O3
InChI KeyRRQYJINTUHWNHW-UHFFFAOYSA-N
SMILESCCOCCOCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (K.F.)
Color scale=<15 APHA
Peroxides=<0.0015 % (as H2O2)
Infrared spectrumConforms
Acidity (CH3COOH)=<0.015 %
View more
Lewis acid
RUO – Research Use Only