Check Order Status
Sodium chloranilate trihydrate, 95%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
Sodium chloranilate trihydrate, 95%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
ACR419500250
25 g
Glass bottle
Catalog number
ACR419500250
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium chloranilate trihydrate
Name Note
95%
CAS
36275-66-8
Color
Brown
EINECS Number
252-947-6