Silver phosphate, 98%
Silver phosphate, 98%
Silver phosphate, 98%
Silver phosphate, 98%
Thermo Scientific Chemicals
Silver phosphate, 98%
CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR41937025025 gGlass bottle
Catalog number ACR419370250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS627-27-0
IUPAC Namebut-3-en-1-ol
Molecular FormulaC4H8O
InChI KeyZSPTYLOMNJNZNG-UHFFFAOYSA-N
SMILESOCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %