Lead citrate, trihydrate, 96%, Thermo Scientific™
Lead citrate, trihydrate, 96%, Thermo Scientific™
Lead citrate, trihydrate, 96%, Thermo Scientific™
Lead citrate, trihydrate, 96%, Thermo Scientific™
Lead citrate, trihydrate, 96%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR413061000100 gGlass bottle
Catalog number ACR413061000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLead citrate, trihydrate
Name Note96%
CAS6107-83-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
Fatal if inhaled. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. May cause damage to organs through prolonged or repeated exposure. May damage the unborn child. Suspected of damaging fertility.
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.