Hydriodic acid, ACS reagent, 55% in water, without stabilizer, Thermo Scientific Chemicals
Hydriodic acid, ACS reagent, 55% in water, without stabilizer, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydriodic acid, ACS reagent, 55% in water, without stabilizer, Thermo Scientific Chemicals
CAS: 10034-85-2 | HI | 127.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Catalog number ACR411840500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydriodic acid
CAS10034-85-2
Boiling Point127.0°C
Health Hazard 1GHS Signal Word: Danger
Density1.7000 g/mL