Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Thermo Scientific Chemicals
Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol
Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant.
Have Questions?
Catalog NumberQuantityPackaging
ACR410731000100 gPlastic bottle
Catalog number ACR410731000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Composition40 to 60 wt% water
FormPowder
Assay (unspecified)nitrobenzene activity: 600, minimum
Assay (unspecified)Pd 4.80 to 5.10 wt% ( dry basis)
General Description
  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol
Applications
  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
RUO – Research Use Only