Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR404410050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroacetic acid sodium salt
Name Note98%
CAS3926-62-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Very toxic to aquatic life.