Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR404410025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS87-59-2
IUPAC Name2,3-dimethylaniline
Molecular FormulaC8H11N
InChI KeyVVAKEQGKZNKUSU-UHFFFAOYSA-N
SMILESCC1=CC=CC(N)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or orange or red-brown or yellow-brown
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.5660-1.5700 @ 20?C