Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR404035000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total trace metal impurities=<0.001 %
Appearance (Form)Crystals and/or chunks
Filter aid
RUO – Research Use Only