Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number ACR404030025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd ( wt %) 9 - 11
Loss on DryingWater (%) 45 - 65
Filter aid
RUO – Research Use Only