Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Thermo Scientific Chemicals
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
CAS: 38402-02-7 | C6H16ClNO2 | 169.65 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR40253025025 gGlass bottle
Catalog number ACR402530250
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1074-24-4
IUPAC Name1,4-dibromo-2,5-dimethylbenzene
Molecular FormulaC8H8Br2
InChI KeyQENIALCDPFDFHX-UHFFFAOYSA-N
SMILESCC1=CC(Br)=C(C)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)70.0-76.0°C (non-U.S. specification)
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms (non-U.S. specification)
Phase transfer catalyst
RUO – Research Use Only