Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Thermo Scientific Chemicals
Biebrich Scarlet, dye content approx. 70%
CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol
Have Questions?
Catalog NumberQuantity
ACR402221000100 g
Catalog number ACR402221000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS92-88-6
IUPAC Name[1,1'-biphenyl]-4,4'-diol
Molecular FormulaC12H10O2
InChI KeyVCCBEIPGXKNHFW-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point278°C to 283°C
Appearance (Color)Off-white to light beige
Infrared spectrumConforms
Appearance (Form)Powder
GC>=96.0 %