Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR401521000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3173-56-6
IUPAC Name(isocyanatomethyl)benzene
Molecular FormulaC8H7NO
InChI KeyYDNLNVZZTACNJX-UHFFFAOYSA-N
SMILESO=C=NCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5241 to 1.5261 (20°C, 589 nm)
GC>=99.0 %