Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR401520250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS138642-62-3
IUPAC Name(2-cyanophenyl)boronic acid
Molecular FormulaC7H6BNO2
InChI KeyNPLZNDDFVCGRAG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)Off-white to cream
HPLC>=97.5 %