1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR3982600505 gGlass bottle
Catalog number ACR398260050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-81-3
IUPAC Name2-bromopentane
Molecular FormulaC5H11Br
InChI KeyLGAJYTCRJPCZRJ-UHFFFAOYNA-N
SMILESCCCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4393 to 1.4413 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<100 APHA
Infrared spectrumConforms
GC>=88 % Typical