(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR397621000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(Trimethylsilyl)diazomethane
Name NoteApprox. 2M solution in diethyl ether
CAS18107-18-1
CAS Min %28.19
Health Hazard 1GHS Signal Word: Danger
View more
Alkylation reagent
RUO – Research Use Only