Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
Catalog number ACR393322500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1188-14-3
IUPAC Nametriethylgermane
Molecular FormulaC6H16Ge
InChI KeyLTLGQZPDSQYFSB-UHFFFAOYSA-N
SMILESCC[GeH](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥96.0%
Suitable for battery materials development.
For Manufacturing and Laboratory Use. Not for beverage use