2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR392790050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13389-42-9
IUPAC Name(2E)-oct-2-ene
Molecular FormulaC8H16
InChI KeyILPBINAXDRFYPL-HWKANZROSA-N
SMILESCCCCCC=CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4100-1.4160 @ 20?C