tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number ACR390690010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Residue on Evaporation5ppm
Optical Absorbance at400nm: 0.01 AU, max
Assay (GC)≥98.0%
Water Content (Karl Fischer Titration)0.02%, max