Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR390070025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS10361-82-7
IUPAC Namesamarium(3+) trichloride
Molecular FormulaCl3Sm
InChI KeyBHXBZLPMVFUQBQ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.