Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
CAS: 1332-09-8 | C28H37FO7 | 509.626 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3900400101 kgPlastic Bottle
Catalog number ACR390040010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS626-55-1
IUPAC Name3-bromopyridine
Molecular FormulaC5H4BrN
InChI KeyNYPYPOZNGOXYSU-UHFFFAOYSA-N
SMILESBrC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5680 to 1.5720 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to light brown
GC>=98.5 %
Appearance (Form)Liquid