Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR389025000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51779-32-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder