Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR383042500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS15060-55-6
IUPAC Namediammonium disulfanylidenemolybdenum disulfanide
Molecular FormulaH10MoN2S4
InChI KeyZKKLPDLKUGTPME-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Molybdenum (Mo)35.5 to 38.1 %
Total trace metal impurities=<500 ppm
Appearance (Color)Dark red or green or purple or black