N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals
CAS: 37595-74-7 | C8H5F6NO4S2 | 357.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR374790050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Phenylbis(trifluoromethanesulfonimide)
CAS37595-74-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only