Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR364335000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS49805-30-3
IUPAC Name2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular FormulaC6H7NO
InChI KeyDDUFYKNOXPZZIW-UHFFFAOYNA-N
SMILESO=C1NC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point55°C to 59°C
GC>=97.5 %