Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
CAS: 31430-18-9 | C14H11N3O3S | 301.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Packaging:
Glass bottle
Catalog number ACR358240500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7252-83-7
IUPAC Name2-bromo-1,1-dimethoxyethane
Molecular FormulaC4H9BrO2
InChI KeyFUSFWUFSEJXMRQ-UHFFFAOYSA-N
SMILESCOC(CBr)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Stabilizer0.2 % potassium carbonate