Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
CAS: 31430-18-9 | C14H11N3O3S | 301.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
Catalog number ACR358240100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNocodazole
Name Note98%
CAS31430-18-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning