Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR349962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9011-14-7
Molecular Formula(C5H8O2)n
SMILESCOC(=O)C(C)(-*)C-*
MDL NumberMFCD00134349
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Appearance (Color)White
Additional infobead size (typically): 10 to 100 micron
Loss on drying=<3 %
Infrared spectrumConforms
Oxidation reagent
RUO – Research Use Only