Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number ACR348630025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPerchloric acid
ColorColorless
Name Note0.1 M Solution in Acetic Acid
CAS7601-90-3
Health Hazard 1Danger
View more
Oxidation reagent
RUO – Research Use Only