Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number ACR348630010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS99-07-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown to dark brown to purple to dark purple
FormPowder
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Oxidation reagent
RUO – Research Use Only