Check Order Status
(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Packaging:
Glass bottle
Catalog number
ACR338820010
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine
Name Note
99%
CAS
80548-31-8
CAS Min %
98.5
Health Hazard 1
GHS Signal Word:
Warning