Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number ACR328271000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7759-02-6
IUPAC Namestrontium(2+) sulfate
Molecular FormulaO4SSr
InChI KeyUBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES[Sr++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.5% max