Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR326590010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-12-0
IUPAC Namedisodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
Molecular FormulaCr2H4Na2O9
InChI KeyJYDRNIYTFCBIFC-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5-100.5% Cr₂Na₂O₇.2H2O
Impurity contentCa 0.003%
Impurity contentK 0.01%
Impurity contentSO₄ 0.01%
Impurity contentCl 0.005%
View more
For residue analysis
RUO – Research Use Only