View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialUrethane
Name Note97%
CAS51-79-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Harmful if swallowed. May cause cancer.