Cacodylic acid, 98%, Thermo Scientific Chemicals
Cacodylic acid, 98%, Thermo Scientific Chemicals
Cacodylic acid, 98%, Thermo Scientific Chemicals
Cacodylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cacodylic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number ACR318150050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCacodylic acid
CAS75-60-5
Melting Point195°C to 198°C
Health Hazard 1GHS Signal Word: Danger