1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 2708-97-6 | C6F4I2 | 401.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR311690050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS99-07-0
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Powder
Appearance (Color)Grey to brown to purple