1,3-Bis(diphenylphosphino)propane, 97%, Thermo Scientific Chemicals
1,3-Bis(diphenylphosphino)propane, 97%, Thermo Scientific Chemicals
1,3-Bis(diphenylphosphino)propane, 97%, Thermo Scientific Chemicals
1,3-Bis(diphenylphosphino)propane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Bis(diphenylphosphino)propane, 97%, Thermo Scientific Chemicals
CAS: 6737-42-4 | C27H26P2 | 412.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR310050050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS261-31-4
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
Melting point127°C to 131°C
Appearance (Form)Crystals or needles