Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number ACR307050100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1313-97-9
IUPAC Namedineodymium(3+) trioxidandiide
Molecular FormulaNd2O3
InChI KeyPLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Nd+3].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.001% max.