Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ACR301221000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS458-37-7
IUPAC Name(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
Molecular FormulaC21H20O6
InChI KeyZIUSSTSXXLLKKK-KOBPDPAPSA-N
SMILESCOC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange-yellow or orange
FormPowder or crystalline powder
Elemental AnalysisC: 68.47 ? 4%
Elemental AnalysisH: 5.47 ? 0.3%